MELLI S1P ESD Safety Sandals for Ladies, Grey
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
741341/35 741341/36 741341/37 741341/38 741341/39 741341/40 741341/41 741341/42
ELTN741341/35EA 679 DKK
ELTN741341/35 ELTN741341/36 ELTN741341/37 ELTN741341/38 ELTN741341/39 ELTN741341/40 ELTN741341/41 ELTN741341/42
MELLI S1P ESD Safety Sandals for Ladies, Grey
Sko og støvler
Ladies' safety sandal, MELLI, Velcro® closure in grey finish.
  • Microfibre/mesh material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole SPORTIVE ESD grey
  • Non-metallic, penetration-resistant midsole
  • TPU/PU sole MAXXIMO
  • Steel toe cap
For use in dry work areas, applications include industry, storage, transport, assembly etc. For areas where there is a risk of penetration from pointed and sharp objects (S1P), or areas where there is a risk of electrostatic discharge (ESDS/ESD). For workplaces on hard undergrounds, the revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides, resulting in more energy in every step.
Certificeringer: EN ISO 20345 S1P SRC, form A.