NELSON XXG GTX S3 HI CI ESD Safety Shoes, Black, Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
728691/39 728691/40 728691/41 728691/42 728691/43 728691/44 728691/45 728691/46 728691/47 728691/48
ELTN728691/39EA 932 DKK
ELTN728691/39 ELTN728691/40 ELTN728691/41 ELTN728691/42 ELTN728691/43 ELTN728691/44 ELTN728691/45 ELTN728691/46 ELTN728691/47 ELTN728691/48
NELSON XXG GTX S3 HI CI ESD Safety Shoes, Black, Low
Sko og støvler
Safety shoe, NELSON XXG GTX, lace-up in black finish.
  • Hydrophobized nubuck leather/hydrophobized textile material
  • GORE-TEX performance comfort footwear
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • Rubber/PU sole WELLMAXX GRIP
  • Composite toe cap
For indoor and outdoor use, in areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), or areas where there is a risk of electrostatic discharge (ESDS/ESD). For workplaces on hard undergrounds, the revolutionary Infinergy® sole core cushions impacts and provides a rebound effect when the compressive impulse subsides, resulting in more energy in every step.
Certificeringer: EN ISO 20345 S3 HI/CI/HRO/SRC, form A.