Pharma Scoops
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8860 8861 8862 8863
BOCH8860EA 1590 DKK
BOCH8860 BOCH8861 BOCH8862 BOCH8863
Pharma Scoops
Øser
These scoops are made from V4A steel.
  • Very good chemical resistance
  • Excellent thermal resistance, autoclavable
  • Highly polished
  • With seamless fixed handle
Ideal for sample taking in the pharmaceutical industry.