Petri Dishes, Glass
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NF632492002040 NF632492002050 NF632492002060 NF632492003060 NF632492007070 NF632492003080 NF632492003090 NF632492001100 NF632492003100 NF632492004100 NF632492004120 NF632492005150 NF632492005180 NF632492006200
KAVANF632492002040EA 4870 DKK
KAVANF632492002040 KAVANF632492002050 KAVANF632492002060 KAVANF632492003060 KAVANF632492007070 KAVANF632492003080 KAVANF632492003090 KAVANF632492001100 KAVANF632492003100 KAVANF632492004100 KAVANF632492004120 KAVANF632492005150 KAVANF632492005180 KAVANF632492006200
Petri Dishes, Glass
Dyrkningsplader Petriskåle
Soda-lime glass, with lid. A green alternative to disposable Petri dishes made from plastic.
  • Good chemical resistance
  • Heatable up to 60 °C
  • Single compartment
  • Without vents
These dishes are not suitable for applications related to strong heat or rapid temperature changes.