Watch Glass Dishes
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NF634951430840 NF634951430770 NF634951430780 NF634951430790 NF634951430860 NF634951430910 NF634951430950 NF634951430010 NF634951430060 NF634951430070 NF634951430080 NF634951430110 NF634952430120 NF634952430160 NF634952430180 NF634952430200 NF634952430250
KAVANF634951430840EA 227 DKK
KAVANF634951430840 KAVANF634951430770 KAVANF634951430780 KAVANF634951430790 KAVANF634951430860 KAVANF634951430910 KAVANF634951430950 KAVANF634951430010 KAVANF634951430060 KAVANF634951430070 KAVANF634951430080 KAVANF634951430110 KAVANF634952430120 KAVANF634952430160 KAVANF634952430180 KAVANF634952430200 KAVANF634952430250
Watch Glass Dishes
Urglas
Watch glass dishes with fused edges made from soda-lime glass.
  • Good chemical and physical properties
  • Quick and easy to clean
Not suitable for applications related to strong heat or rapid temperature changes.
Certificeringer: DIN 12341.