Firefly luciferase flash assay kit, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16174 16175
PIER16174EA 991 DKK
PIER16174 PIER16175
Firefly luciferase flash assay kit, Pierce™
Assays Proteinassays
The Pierce™ Firefly Luciferase Flash Assay Kit provides reagents to measure red firefly luciferase activity in whole cell producing a very strong flash signal which decays rapidly.
  • Highly sensitive detection of firefly luciferase activity
  • Amenable to automation
  • Contains a universal cell lysis buffer and optimized flash assay reagent
The bioluminescent signal produced by firefly luciferase results from the oxidation of D-Luciferin. The light output correlates with the amount of firefly protein expressed, which is proportional to the activity of the promoter for firefly expression.
Leveringsinformation: Kit contains Firefly Flash Assay Buffer (5 or 50 mL), D-Luciferin (3 or 30 mg) and 2X Cell Lysis Buffer (6 or 60 mL).
Advarsel: For research use only. Not for use in diagnostic procedures. It stored at 4 °C.