Renilla luciferase glow assay kits, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16166 16167
PIER16166EA 1150 DKK
PIER16166 PIER16167
Renilla luciferase glow assay kits, Pierce™
Assays Enzym Assays
The Thermo Scientific Pierce renilla Luciferase glow assay kit provides an extremely bright and stable bioluminescence signal (half-life approx. 3 hours), especially in the presence of green renilla luciferase reporter.
  • Sensitive - highly sensitive detection of Green Renilla luciferase activity
  • Stable - increased signal stability compared to flash assays
  • Compatible - assay reagents compatible with other Renilla luciferases
  • Automation-friendly - amenable to high throughput assays
  • Convenient - contains a universal cell lysis buffer and optimized glow assay reagent
Leveringsinformation: Kit contains Renilla glow assay buffer (5 ml); 100X coelenterazine (50 µl); and 2X cell lysis buffer (6 ml).
Advarsel: For research use only. Not for use in diagnostic procedures.