Crucibles, stainless steel
Leverandør: USBECK Laborgeräte
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5640 5641 5642 5643
USBE5640EA 164 DKK
USBE5640 USBE5641 USBE5642 USBE5643
Crucibles, stainless steel
Digler
Crucibles without lid made from stainless steel 18/10.
  • Highly resistant to chemicals and reagents
  • Withstand maximum temperature of 800 °C
  • Autoclavable
Bestillingsoplysninger: Lids need to be ordered separately.