Protein Coated Cell Culture Dishes, CELLCOAT®, Collagen Type I
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
628950_100 664950_40
392-0271EA 4510 DKK
392-0271 392-0279
Protein Coated Cell Culture Dishes, CELLCOAT®, Collagen Type I
Dyrkningsplader Celledyrkningsskåle
PS, coated with rat tail collagen type I. Collagen type I is an extracellular matrix protein, which in vivo influences adhesion, migration and proliferation, amongst other processes. Many otherwise difficult-to-cultivate cells adhere to collagen type I and show positive growth behaviour. For certain cell lines collagen type I also has an influence on differentiation and morphology.
  • Improved cell adhesion, proliferation and growth of endothelial cells, hepatocytes, muscle cells, pheochromocytoma cells (PC12), as well as other cell types
  • Suitable for serum-free or serum-reduced cell cultivation
  • Quality control tested to demonstrate promotion of the adhesion and proliferation of human fibrocarcoma cells
Shelf life: 24 months at room temperature