VWR®, XL Extended Length Pipette Tips, 200 µl
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
613-7088EA 232 DKK
613-7088 732-3908
VWR®, XL Extended Length Pipette Tips, 200 µl
Pipettespidser
Easily reach the bottom of 12×75 mm tubes without contaminating the pipettor nose cone.
  • Extended length (91 mm)
  • Universal fit for compatibility with most major pipettor brands
  • Low-retention resin technology creates a low-binding surface for maximum sample recovery
  • Graduations at 10, 50, 100, 150, and 200 μl
  • 96-tip format in a hinged, locking rack that easily accommodates multi-channel pipettors
Filter placement is optimised to accommodate 20, 50, 100, and 200 μl pipettors; non-filtered tips are also compatible with 300 μl pipettors, eliminating the need to keep multiple tips in inventory.
Certificeringer: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.