Smiths Medical® Luer Stopcock Fittings, Polycarbonate
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX30600-00BLKEA 5740 DKK
MFLX30600-00BLK MFLX30600-03BLK MFLX30600-04BLK MFLX30600-02BLK MFLX30600-01BLK
Smiths Medical® Luer Stopcock Fittings, Polycarbonate
Ventiler Kugleventiler
Regulate flow without any leaking.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.