PCR plates, 96-Well, rigid frame, PC/PP, BIO-CERT® PCR QUALITY
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
781541 781542 781543 781544 781545 781546 781547 781548 781549
732-3918EA 2730 DKK
732-3918 732-3919 732-3920 732-3921 732-3922 732-3923 732-3924 732-3925 732-3926
PCR plates, 96-Well, rigid frame, PC/PP, BIO-CERT® PCR QUALITY
Mikroplader PCR plader
For medium and high sample throughput, these 96-well plates are the efficient solution for PCR and qPCR.
  • High stability and rigidity of the plate due to the polycarbonate frame
  • Rapid heat transfer through thin vessel walls
  • Suitable for all standard thermal cyclers and fully automation compatible
The rigid frame PCR plates have a strong polycarbonate frame to ensure safe handling in semiautomated and automated pipetting systems and optimal evaporation protection. The rigid frame also allows the use of very thin-walled wells from 100% virgin medical grade polypropylene. The uniform and thin wall thicknesses of the wells ensure rapid transfer of the temperature from the cycler to the sample, thus reducing cycle times.
A number of skirt choices ensures a good fit in common thermal cyclers for efficient heat transfer. Choose between semi-skirted and fully skirted plates. Different colours help to distinguish between the workflow steps. For qPCR, white plates are available that optimally reflect the fluorescence signals. The cleanroom quality of the PCR consumables from BRAND, guarantees reliable results. BRAND 96-well Rigid Frame plates are produced under controlled cleanroom conditions and thus meet the purity criteria of the quality level BIO-CERT® PCR QUALITY.