ISO AIR cleanroom masks with tie-on closures
Leverandør: Medicom
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60.720L 60.720M
114-9502EA 1040 DKK
114-9502 114-9501
ISO AIR cleanroom masks with tie-on closures
Masker Renrumsmasker
ISO AIR cleanroom masks with tie-on closures provide optimal comfort thanks to soft, non-irritating elastics.
  • Made in France
  • Made from 30% recycled polypropylene material
  • High breathability
  • ISO 3 Grade B protection
Masks feature low particle release, and are Helmke drum tested 'excellent' for filtration efficiency (BFE, PFE, VFE >99%). Softex technology limits the risk of particulate contamination while reducing the risk of skin irritation. An adjustable and malleable nose clip prevents glasses from fogging up. Tie-on masks are simple to use, with long ties that allow easy installation and fitting.
Certificeringer: The ISO AIR range is manufactured in an ISO 8 cleanroom.
Emballering: 6 double packed bags × 50 units.