ISO AIR cleanroom masks with earloops
Leverandør: Medicom
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60.712L 60.712M
114-9498EA 799 DKK
114-9498 114-9497
ISO AIR cleanroom masks with earloops
Masker Renrumsmasker
ISO AIR cleanroom masks with earloops provide optimal comfort thanks to soft, non-irritating elastics.
  • Made in France
  • Made from 25% recycled polypropylene material
  • High breathability
  • ISO 3 Grade B protection
Masks feature low particle release, and are Helmke drum tested 'excellent' for filtration efficiency (BFE, PFE, VFE >99%). Softex technology limits the risk of particulate contamination while reducing the risk of skin irritation. An adjustable and malleable nose clip prevents glasses from fogging up.
Certificeringer: The ISO AIR range isMore Product Information