Polyester Film Tapes, 850
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
850R50 850T19 850T2566 850S5166
MMMA850R50EA 9480 DKK
MMMA850R50 MMMA850T19 MMMA850T2566 MMMA850S5166
Polyester Film Tapes, 850
Tape
3M™ polyester tape 850 is designed for splicing applications requiring quick stick and an aggressive, permanent hold. Used in a variety of applications, including web processing, label protection, and marking.
  • Aggressive acrylic adhesive provides quick stick and excellent long-term holding in splicing applications
  • Thin, high-strength polyester film meets low calliper requirements and will not interfere with most machinery processes
  • Polyester backing resists tearing and protects surfaces from abrasion and rough handling
  • Temperature resistance enables tape to perform in high temperature operations
3M™ polyester film tape 850 is resistant to chemicals and solvents, helping to reduce damage and increase output. It offers excellent tear and temperature resistance, thus increasing durability. It also provides abrasion resistance that protects its surface from rough handling. It features a pressure sensitive acrylic adhesive for clarity and long-term holding. Available in a variety of colours, including a transparent option that blends well with web stock.