Hard hats, G3000 series
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G30DUY G31NUW G31MUOIS
111-2314EA 302 DKK
111-2314 MMMAG31NUW MMMAG31MUOIS
Hard hats, G3000 series
Hjelme Sikkerhedshjelme
The 3M™ G3000 series hard hats are designed in close collaboration with industrial and forestry workers. They are intended for use in harsh environments with tough demands for effective protection, excellent ventilation and a maximum field of vision.
  • Designed for use in harsh environments, with excellent ventilation and a good field of vision
  • 3M™ Uvicator™ sensor disc indicates overexposure to UV-radiation and appropriate replacement time
  • Softly rounded design helps avoid parts snagging on cables or branches
  • Ventilation holes distributed over a large area offer maximum through-flow of air
  • Head harness can be rotated 180° for better visibility when working in confined spaces
  • Short peak provides improved upwards visibility
  • Replaceable leather or plastic sweatband
  • Approved for use at −30 °C
3M™ G3000 series hard hats are available with pinlock or ratchet suspension and a plastic or leather sweatband. It’s available with or without ventilation holes that are distributed over a large area offer maximum through-flow of air, increasing wearer comfort.
The slim design means that falling objects slide off easily and do not dislodge the helmet from its position. Softly rounded design, without any sharp or protruding components, the extremely comfortable lining and the simple adjustment of the neck strap make the G3000 series unique.
Certificeringer: Helmets meet EN 397:212+A1:2012 requirements with the harness rotated through 180° (back to front). Electrically insulated versions meet EN 50365:2002 for use on low voltage installations.