PURELL® Advanced Hygienic Hand Rub Bottles
Leverandør: GOJO Industries - Europe Ltd
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9661-24EEU00 9663-12
GOJO9661-24EEU00EA 34.2 DKK
GOJO9661-24EEU00 GOJO9663-12
PURELL® Advanced Hygienic Hand Rub Bottles
Hudpleje Sæber og huden rensemidler
A no-rinse hygienic hand rub that feels great to use and is clinically proven to maintain skin health.
  • Includes moisturisers to help keep skin healthy
  • Kills 99.99% of most common germs that may be harmful
  • 70% alcohol formulation is suitable for frequent use by hospital staff
  • Recommended for use on hands that are lightly soiled and in conjunction with a hand hygiene regime
  • Halal certified
Bactericidal according to EN 1500 (hand sanitiser). Conforms to EN 12791 (surgical rub), EN 1276, and EN 13727, virucidal according to EN 14476, fungicidal and yeasticidal according to EN 13624 and EN 1650; and mycobactericidal according to EN 14348. Food taint test compliant: BS EN 4120:2007.