Chloromethyl chlorosulphate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Chloromethyl chlorosulphonate, Chlorosulphuric acid Chloromethyl Ester, Chlorosulphonic acid Chloromethyl Ester

OR13650-5G OR13650-100G OR13650-25G
APOSOR13650-5GEA 0 DKK
APOSOR13650-5G APOSOR13650-100G APOSOR13650-25G
Chloromethyl chlorosulphate
Chloromethyl chlorosulphonate
Formel: CH₂Cl₂O₃S
Kogepunkt: 45-50/10mm °C
Massefylde: 1,631 g/cm³
MDL Number: MFCD00041547
CAS-nummer: 49715-04-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR