Four drawer storage cupboards
Leverandør: TEKNOMEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SHCBED10X60X20 SHCBED12X60X20 SHCBID10X60X20 SHCBID12X60X20
TEKMSHCBED10X60X20EA 0 DKK
TEKMSHCBED10X60X20 TEKMSHCBED12X60X20 TEKMSHCBID10X60X20 TEKMSHCBID12X60X20
Four drawer storage cupboards
Møbler Opbevaringskabinetter
Our storage cupboard comes complete with four drawers to provide perfect storage.
  • Brushed dull polish finish
  • Sloping top
  • Four shelf height adjustments
  • Stainless steel adjustable feet with non-marking rubber base
  • Lockable main doors
  • Four internal or external drawers
  • Shelves sold separately
Our cupboard has been designed with a sloping top, four drawers (internal and external options available) and lockable doors to secure all content.
The range includes a single (600 mm) or double (1000 or 1200 mm) solid door.
Supplied with four easy clean shelf height adjustments, just add the number of shelves necessary to suit your needs.
Our cupboards are tough, with a robust 1,5 mm thick stainless steel, they are designed with hygiene in mind. No ledges or crevices assures that dirt cannot find anywhere to hide, while the solid stainless steel construction enables easy cleaning. Further, the 300 mm legs allows easy access to any dirt hiding beneath and audit inspections.