PCR tubes and caps, Axygen®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PCR-02-A PCR-02-C PCR-02-L-C PCR-05-A PCR-05-C PCR-05-L-C PCR-02-C-500 PCR-05-C-500
732-1033EA 528 DKK
732-1033 732-0674 525-0249 732-1035 732-0675 525-0251 731-0679 731-0680
PCR tubes and caps, Axygen®
Rør PCR rør
Axygen thin wall PCR strip tubes are designed to fit a variety of thermal cyclers. The consistent and ultra-thin wall allows precise thermal transfer.
An even insertion cap ensures a tight fit to reduce evaporation. Strip tubes are available in an assortment of colors.
Optical flat cap strips feature ultra-clear upper surfaces, ideal for fluorescent luminescent and real-time PCR applications.
They offer a light transmittance that is 75% higher than with standard strip caps. Made to seal tube strips and 96-well microplates.