Industrial gloves, oil repellent, EDGE® 48-501
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
48-501/10 48-501/8 48-501/9
ANSE48-501/10EA 325 DKK
ANSE48-501/10 ANSE48-501/8 ANSE48-501/9
Industrial gloves, oil repellent, EDGE® 48-501
Handsker
Heavy-duty work gloves with cotton liner, providing good defense against oils, liquids and abrasion.
  • Good level of abrasion protection
  • Nitrile coating (3/4) delivers long-lasting performance
  • Oil and liquid repellent
  • FDA-compliant materials: Suitable for food contact and packaging
A cost-effective solution for heavy weight applications requiring a good level of abrasion resistance, along with liquid repellency. Nitrile dipped cut and sew glove for heavy handling, offering good resistance to abrasive material. Stronger and more flexible than PVC, while providing a good barrier to grease and oil. Will not degrade like leather or latex. The economical choice for a heavy-duty glove with quality features.
Recommended for: Cold plastic moldings; heavy-duty cables; rough casting and cores; steel bars, sheet metals, cans and other metal products; street cleaning and refuse collection.
Certificeringer: EN 388:2016 (4111B), Cat. II