VWR®, Microcentrifuge Tubes, Low-Binding
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525-1492EA 274 DKK
525-1492 525-1493 525-1601
VWR®, Microcentrifuge Tubes, Low-Binding
Rør Mikrocentrifugerør
These microcentrifuge tubes have a low adhesion surface which provides minimal to low binding. For use with DNA and viscous proteins, these non stick, graduated, polypropylene tubes may be used between –80 to +122 °C (–122 to +251,6 °F).
  • Leak proof snap caps
  • Autoclavable
  • Graduated markings every 100 µl
  • Rated to 20000×g
  • All tubes are nonsterile
  • Certified free of RNase, DNase and pyrogens
VWR® microcentrifuge tubes feature flat caps and frosted areas on the sides for easy labeling. Leakproof snap caps may be penetrated by a syringe. Easy-to-open caps will not pop open during autoclaving. The 0,5 ml tubes fit many thermal cyclers.