24-well permeable supports system, polystyrene, sterile, HTS Transwell®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3396 3397 3398 3399 3378 3379
734-1559EA 4370 DKK
734-1559 734-1560 734-1561 734-1562 734-4072 734-4073
24-well permeable supports system, polystyrene, sterile, HTS Transwell®
Mikroplader HTS mikroplader
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
  • Cell growth area is 0,33 cm²/well
  • Choice of either polyester (PET) membrane (0,4 µm pore size) or polycarbonate (PC) membrane (0,49 m and 3,0 µm pore sizes)
  • Treated for optimal cell attachment
  • Sterile
The HTS Transwell-24 permeable support has an array of 24 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 24 Transwell supports to be handled as a single unit.
Bestillingsoplysninger: Individual pack has two HTS Transwell-24 units loaded into two open reservoir trays and two individually wrapped 24-well plates. Bulk pack has 12 HTS Transwell-24 units loaded into 24-well plates only. Reservoirs may be purchased separately.