Welding gloves, EDGE® 48-216
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
48-216/10 48-216/11
ANSE48-216/10EA 639 DKK
ANSE48-216/10 ANSE48-216/11
Welding gloves, EDGE® 48-216
Handsker
These protective gloves offer EN 12477 Type A protection for demanding welding and thermal job tasks, ranging from stick and MIG welding to torch cutting and grinding.
  • Improved three-dimensional thumb design
  • High-dexterity gloves offer tactile sensitivity for precise welding control
  • Long cuff extends protection over the forearm
  • Higher durability as they are made of premium split cowhide leather and have welted seams sewn with aramid
  • Breathable cotton lining grants thermal insulation and extended comfort
Recommended for: Brazing, handling hot metal castings, grinding, welding, handling of metal sheets, cutting of dry, painted or galvanised pieces.
Certificeringer: EN 388:2016 (4133B), EN 407 (413X4X), EN 12477:2001 + A1:2005; Cat. III