Industrial nitrile gloves, AlphaTec® 37-300 series
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
37-300/10 37-300/11 37-300/7 37-300/8 37-300/9
ANSE37-300/10EA 133 DKK
ANSE37-300/10 ANSE37-300/11 ANSE37-300/7 ANSE37-300/8 ANSE37-300/9
Industrial nitrile gloves, AlphaTec® 37-300 series
Handsker
Improved grip, dexterity, and flexibility.
  • 8 mil/0,2 mm thickness allows dexterity and tactility without a loss of durability
  • NBR offering protection against light acids, greases, oils, and liquids
  • Long cuff extends to the forearm
These 0,20 mm thick nitrile gloves are engineeredto provide an optimum balance of fit and flexibility, either on their own or as over gloves. Latex-free formulation presents no risk of Type 1 allergies.