Industrial gloves, HyFlex® 11-944
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-944/10 11-944/7 11-944/8 11-944/9
ANSE11-944/10EA 370 DKK
ANSE11-944/10 ANSE11-944/7 ANSE11-944/8 ANSE11-944/9
Industrial gloves, HyFlex® 11-944
Handsker
Designed for use in moderately oily manufacturing and maintenance environments.
  • Extreme durability
  • Optimal dry grip
  • Great flexibility and dexterity
An ideal alternative to light leather gloves, with fine and stretchable stitches for excellent comfort. Designed specifically for use in moderately oily manufacturing and maintenance environments where protection combined with abrasion resistance are essential.
Certificeringer: EN 388:2016 (3121A); Cat. II