Cut-resistant industrial gloves, HyFlex® 11-739 series
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11-739/10 11-739/11 11-739/6 11-739/7 11-739/8 11-739/9
ANSE11-739/10EA 2430 DKK
ANSE11-739/10 ANSE11-739/11 ANSE11-739/6 ANSE11-739/7 ANSE11-739/8 ANSE11-739/9
Cut-resistant industrial gloves, HyFlex® 11-739 series
Handsker
Dexterity with superb cut resistance for optimal productivity.
  • Offering good levels of cut resistance
  • PU coating provides excellent grip
The glove liner provides good levels of cut resistance for safe use in controlling, handling, and assembly applications involving contact with sharp pieces. The specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces. The PU formulation also increases abrasion resistance on the glove's work surface.