Perforated tweezers, Swiss Tweeze®
Leverandør: OUTILS RUBIS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5ARG 4G 1G 2AG 2G 3CG 3G 5AG 5G 7G
232-0231EA 257 DKK
232-0231 232-0222 232-0223 232-0225 232-0224 232-0227 232-0226 232-0229 232-0228 232-0230
Perforated tweezers, Swiss Tweeze®
Pincet
Perforated tweezers engineered for a non slip grip makes them ultra-light, granting superior handling and complete control.
  • Made from anti-magnetic stainless steel
  • Enhanced grip and lighter weight due to perforation
  • Autoclavable
Enhance the grip of the tweezers without applying any coating means that the tweezers can be sterilised as often happens in controlled environment workspaces such as cleanrooms.
Swiss Tweeze® is a registered design and is where design and functionality meet to create state-of-art tweezers.