96-well permeable support systems and plates, HTS Transwell®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3374 3380 3381 3384 3385 3386 3387 3388 3391 3392 7369
734-4071EA 1880 DKK
734-4071 734-4074 734-4075 734-4078 734-4079 734-4080 734-4081 734-4082 734-4083 734-4084 734-3204
96-well permeable support systems and plates, HTS Transwell®
Mikroplader Celledyrkningsplader
Permeable supports for HTS (high throughput screening) applications are excellent for ADMET screening, drug transport, and other assays.
The HTS Transwell® 96-well permeable support features 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit. Available with polyester or polycarbonate membranes in a variety of pore sizes. The 0,143 cm² well membrane area provides significantly more surface area than comparable systems. Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access. Plates are optimised for automation, with multichannel feeder ports, an improved gripping surface, and standard bar codes. The reservoir plate allows for the simultaneous feeding of 96 wells, and is supplied with a removable media stabiliser to minimise the risk of spills. The tissue culture-treated receiver plates isolate each well to enable 96 individual assays. Components are sterilised by gamma radiation.
Bestillingsoplysninger: Support systems consist of the 96-well insert plate packaged with a single-well reservoir plate, as well as a separate 96-well receiver plate for cell growth or assays, and two lids to minimize evaporation and prevent contamination. Support plates consist of the 96-well insert plate packaged with a 96-well receiver plate and one lid (reservoir plate not included).