Low volume microplates 96-well polystyrene, Corning® Stripwell™
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2481 2483 2485
732-5577EA 7050 DKK
732-5577 734-1202 734-1204
Low volume microplates 96-well polystyrene, Corning® Stripwell™
Mikroplader Immunologi mikroplader
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an ‘egg-crate’ style strip holder that allows everyone well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format
  • Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
  • Strips 1×8 are designed to fit only one way into the strip holder, eliminating the chance of misorientation