Gamma Irradiated Sterility Maintenance Covers, Heat Seal
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3212ST 3215ST 3220ST 3222ST 3230ST 3234ST 3235ST
129-3329EA 23210 DKK
129-3329 129-3330 129-3331 129-3332 129-3333 129-3334 129-3335
Gamma Irradiated Sterility Maintenance Covers, Heat Seal
Poser Steriliseringsposer
Ideal for pharmaceutical and biotech production applications.
  • Wide range of sizes
  • Works well with impulse and rotary heat sealers
  • No tools required to open
  • Impervious to liquid sanitisation agents
  • Visibility inside the bag allows for identification
  • Prolong the storage and shelf-life of products sterilised 'in-house'
After unloading the autoclave, under Grade A LAF, place wrapped, steam sterilised items into sterility maintenance bags and seal closed. This technique captures the conditions of Grade A and protects sterilised products from external sources of contamination during storage and transport through lesser classified areas.
Certificeringer: Certificates of Irradiation and Certificates of Conformance sent with shipments.
Emballering: The sterility maintenance covers are packaged in Tyvek Chevron peel pouches with Lot # and expiration date for traceability.