VWR®, Test Tubes without Rim
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
212-0734EA 370 DKK
212-0734 212-0735 212-0736 212-0738 212-0739 212-0740 212-0741 212-0742 212-0737 212-0743
VWR®, Test Tubes without Rim
Rør Reagensglas
Test tubes made from borosilicate 7.0 expansion, ideal for use as disposable.
  • Thick walled
  • Good chemical and physical properties
  • Not suitable for application related to strong heat or rapid temperature changes
An economically proven alternative in an economy size pack.