Corning® Gosselin™ Spreaders
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ETAR-01 ETAT-01 ETAR-05 ETAR-06 ETAR-02 ETAT-03
GOSSETAR-01EA 5500 DKK
GOSSETAR-01 GOSSETAT-01 GOSSETAR-05 GOSSETAR-06 GOSSETAR-02 GOSSETAT-03
Corning® Gosselin™ Spreaders
Spredere Celle Spredere
These spreaders are made of PS.
  • Curved end or "T" end
  • Sterile
They are packed in a tamper-evident resealable bag.