HH85 UHF ACD, RFID Scanners, NFC/UHF, Handheld, with Pistol Grip
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
310383 310384
BRDY310383EA 22840 DKK
BRDY310383 BRDY310384
HH85 UHF ACD, RFID Scanners, NFC/UHF, Handheld, with Pistol Grip
Stregkodesystemer
HH85 RFID ACD scanners are an excellent RFID scanning choice and are a perfect match for Brady SmartID solutions. Available in two versions: With or without LTE.
  • Reads all major 1D barcodes and 2D codes
  • Rugged scanners for data collection in challenging industrial environments
  • Ergonomic design for extended usage IP65 rating and fully drop-tested
  • Perfect RFID legibility for a multitude of RFID labels
  • Extended read range by using di-pole antenna
  • Visual (LED’s) and haptic (vibration) feedback
  • With rubber bumpers and pistol grip for easy handling
  • Battery Hotswap: Replace an empty battery and install a fully charged one in a matter of seconds
The Brady HH83 models are perfect for daily RFID scanning. They enable users to pick up a large range of RFID signals and offer a great user experience.
RFID Capabilities: NFC/HF and UHF
Reading Speed: Up to 1000 tags per second
Focal Point: Up to 15 m
Display Resolution: 1280×720 pixels
Connectivity Options:
Non-LTE model: Bluetooth, USB 2.0, Wireless 802.11 a/b/g/n/ac
LTE model: 3G, 4G, Bluetooth, GPRS/EDGE, USB 2.0, Wireless 802.11 a/b/g/n/ac
Leveringsinformation: Supplied with changeable battery (pre-installed), safety and regulations card, wrist strap.