1,2-Propandiol ≥99.5% USP
Leverandør: Spectrum ChemicalSamlet vurdering
Gennemsntilig vurdering
Synonymer:
Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
Propylene Glycol, USP is used in pharmaceutical products as an excipient solvent in oral and topical drug products. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formel:
CH₃CH(OH)CH₂OH Molekylvægt: 76.1 g/mol Kogepunkt: 187.6 Smeltepunkt: -59 Flammepunkt: 99 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064272 CAS-nummer: 57-55-6 Merck Index: 13,07947 |
|
Specifikationer Testresultater
Assay | 99.5 % min. |
Specific Gravity @ 25°C | 1.035 - 1.037 |
Acidity | To Pass Test |
Water Determination | max. 0.2 % |
Residue On Ignition | max. 0.007 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...