High-capacity toploading laboratory balances, Cubis® II MCE
Leverandør: Sartorius Balances
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MCE32202P2S000 MCE70201S2S000 MCE50201S2S000 MCE36201S2S000 MCE36201P2S000 MCE20201S2S000 MCE70200S2S000 MCE36200S2S000 MCE2202S2S00-0
SARBMCE32202P2S000EA 108400 DKK
SARBMCE32202P2S000 SARBMCE70201S2S000 SARBMCE50201S2S000 SARBMCE36201S2S000 SARBMCE36201P2S000 SARBMCE20201S2S000 SARBMCE70200S2S000 SARBMCE36200S2S000 SARBMCE2202S2S00-0
High-capacity toploading laboratory balances, Cubis® II MCE
Vægte
Cubis® II MCE high-capacity toploading balances with essential user interface offer a maximum load between 11,2 and 70,2 kg and readability between 0,1 and 1 g provide the ideal model for every application.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringentMore Product Information