Caps for 2 ml glass snap top vials, level 2 high-throughput applications, SureSTART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6ARC11NRT1 6ARC11NRT1F 6ARC11NRT1R 6ARC11NRT1RT 6ARC11NRT1T 6ARC11ST1 6ARC11ST1F 6ARC11ST1R 6ARC11ST1RF 6ARC11ST1RT 6ARC11ST1T
HYPE6ARC11NRT1EA 229 DKK
HYPE6ARC11NRT1 HYPE6ARC11NRT1F HYPE6ARC11NRT1R HYPE6ARC11NRT1RT HYPE6ARC11NRT1T HYPE6ARC11ST1 HYPE6ARC11ST1F HYPE6ARC11ST1R HYPE6ARC11ST1RF HYPE6ARC11ST1RT HYPE6ARC11ST1T
Caps for 2 ml glass snap top vials, level 2 high-throughput applications, SureSTART™
Låg Låg til kromatografirør
Choose Thermo Scientific™ SureSTART™ 2 ml glass snap vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems.
Use level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. Available in clear and amber glass, snap top vials have very low metal content, extractables, and leachables and conform to high standards so that you can to meet your regulatory requirements. They are specification certifiedMore Product Information