EPA Certified screw vial and cap kits, level 2 high-throughput applications, SureSTART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6AK12ACT 6AK12CCT 6AK16ACT 6AK16CCT 6AK20ACTAS 6AK20CCTAS 6AK40ACT 6AK40AOTAS 6AK40CCT 6AK40COTAS 6AK4ACT 6AK4CCT 6AK8ACT 6AK8CCT
HYPE6AK12ACTEA 0 DKK
HYPE6AK12ACT HYPE6AK12CCT HYPE6AK16ACT HYPE6AK16CCT HYPE6AK20ACTAS HYPE6AK20CCTAS HYPE6AK40ACT HYPE6AK40AOTAS HYPE6AK40CCT HYPE6AK40COTAS HYPE6AK4ACT HYPE6AK4CCT HYPE6AK8ACT HYPE6AK8CCT
EPA Certified screw vial and cap kits, level 2 high-throughput applications, SureSTART™
Hætteglas
Ensure that your environmental analyses meet government regulations with our level 2 Thermo Scientific™ SureSTART™ EPA certified screw vial and cap kits. Select level 2 products for high throughput applications, when robustness and reproducibility are key.
Choose from a variety of kits containing screw vials in clear or amber glass and open or closed top polypropylene caps with silicone/PTFE or polyethylene foam/PTFE septa designed to reduce autosampler needle issues.