MSCERT+ Screw vial and cap kits, level 3 High performance applications, SureSTART™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6PMCK30LVW 6PMCK341W 6PMCK351W 6PMCK35W 6PMCK36LVW 6PMCK39TR 6PMCK40W 6PMCK41W
548-2768EA 0 DKK
548-2768 HYPE6PMCK341W HYPE6PMCK351W HYPE6PMCK35W 548-2769 HYPE6PMCK39TR HYPE6PMCK40W HYPE6PMCK41W
MSCERT+ Screw vial and cap kits, level 3 High performance applications, SureSTART™
Hætteglas
Meet your certification and documentation needs with our level 3 Thermo Scientific™ SureSTART™ MSCERT+ screw vial and cap kits. Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity.
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass and polypropylene caps with silicone/PTFE septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps for effective seals, consistent data, and increased sample throughput.