Floor mattings, Notrax® 826 Diamond Stat™
Leverandør: JUSTRITE MANUFACTURING
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
826S0023BL 826S0035BL 826S0312BL 826R0375BL 826R0475BL
115-2833EA 1090 DKK
115-2833 115-2834 115-2836 115-2837 115-2838
Floor mattings, Notrax® 826 Diamond Stat™
Måtter Rentrums Måtter til Kontamineringskontrol
Diamond Stat™ is a dissipative/anti-static mat designed to absorb static electricity.
  • Electrically conductive PVC diamond plate top surface
  • Dense closed cell PVC foam base
  • Diamond-plate surface texture provides added traction
  • UniFusion™ bond guaranteed for the life of the mat
  • Redstop™ non-slip backing to reduce mat slippage
  • Bevelled edges on all 4 sides
  • Certified by the NFSI (National Floor Safety Institute)
  • Grounding cord included, required
The mat drains static electricity from the worker, keeping sensitive circuitry from being damaged. The durableMore Product Information