PTFE tapes, ResTape
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22485 22486 22487
REST22485EA 93.3 DKK
REST22485 REST22486 REST22487
PTFE tapes, ResTape
Tape
ResTape PTFE tape is for use with threaded connections in a wide range of plumbing materials.
  • Each roll is 12,7 mm × 6,6 m
Tapes have a maximum effective temperature of 260 °C.