CRP gamma irradiated cleanroom trash bags, 2 mil.
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S3655CP1 S3655CP1DP S3655CP2 S3655CP2DP S3655CP3 S3655CP3DP S3655CP4 S3655CP4DP S3655CP5 S3655CP5DP S3655CP10 S3655CP10DP S2428CP1 S2428CP1DP S2428CP5 S2428CP5DP S2428CP10 S2428CP10DP
129-2939EA 9400 DKK
129-2939 129-2918 129-2917 129-2916 129-2915 129-2914 129-2913 129-2912 129-2907 129-2906 129-2920 129-2919 129-2959 129-2962 129-2963 129-2964 129-2960 129-2961
CRP gamma irradiated cleanroom trash bags, 2 mil.
Poser Affaldsposer
Ready to use pre-sterilised trash bags make it easy to remove waste from pharmaceutical production areas without introducing potential sources of contamination.
  • Gamma irradiated
  • Convenient way to hold refuse or other items for removal from an aseptic production area
These rugged bags are supplied sterile in easy-to-open Tyvek® peel pouches and eliminate the need to sterilise or sanitise waste containers or bags prior to bringing them into the controlled environment. Each pouch includes a lot number for traceability.
Certificeringer: Certificates of irradiation and conformance with each shipment.