4-Nitrobenzenediazonium tetrafluoroborate 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H55315.03 H55315.06 H55315.14
H55315.03EA 427 DKK
H55315.03 H55315.06 H55315.14
4-Nitrobenzenediazonium tetrafluoroborate 97%
4-Nitrobenzenediazonium tetrafluoroborate
Formel: C₆H₄BF₄N₃O₂
Smeltepunkt: 144…148 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012005
CAS-nummer: 456-27-9
EINECS: 207-261-1
UN nummer: 2923
ADR: "8",III

Bestil nu

Specifikationer Testresultater

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR