Liquid and powder bottles, SteriWare
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8404H-1000S 8404H-2000S 8404H-250S
300-0593EA 3520 DKK
300-0593 SAMP8404H-2000S SAMP8404H-250S
Liquid and powder bottles, SteriWare
Flasker Plastflasker
These sterile bottles are ideal for use in laboratories and production areas. The bottles have a wide neck which makes them ideal when working with powders as well as liquids.
  • Wide mouth bottles for easy filling
  • HDPE body with a screw on PP cap
  • Made from FDA conforming polymers
  • Packed in a Cleanroom
  • Individually bagged
  • Sterilised by Gamma
  • BSE/TSE-free
The packaging process takes place in a Class 7 Cleanroom and each bottle is packed into a laminate film bag. After packing the bottles are sterilised by gamma irradiation.