Piperitone ≥90%, liquid
Leverandør: Cayman ChemicalSamlet vurdering
Gennemsntilig vurdering
Synonymer:
6-Isopropyl-3-methyl-2-cyclohexen-1-one predominantly
Piperitone is a monoterpenoid that has been found in Mentha and has diverse biological activities. It is active against E. coli, P. aeruginosa, S. typhimurium, S. aureus, R. leguminosarum, and B. subtilis in agar diffusion assays.
- Diverse biological activities
- Origin: Plant/Ocimum gratissimum
Piperitone enhances the antibacterial activity of nitrofurantoin against several nitrofurantoin-resistant species of Enterobacteriaceae, including C. freundii, E. coli, and S. marcescens, by 3- to 20-fold.2 It exhibits feeding deterrent and repellent activities against ants (Crematogaster) with 80% deterrence index (D80) and 80% repellent index (R80) values of 0.13 µg/µl and 8.9 µg/cm2, respectively. Piperitone is insecticidal against C. maculatus eggs, larvae, and adults.
Advarsel: This product is not for human or veterinary use.
Formel:
C₁₀H₁₆O Molekylvægt: 152.24 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00045532 CAS-nummer: 89-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...