Lanosterol 50%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (3S,5R,10S,13R,14R,17R)-4,4,10,13,14-Pentamethyl-17-((R)-6-methylhept-5-en-2-yl)-2,3,4,5,6,7,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol, Cryptosterol

466720050 466720250
ACRO466720050EA 703 DKK
ACRO466720050 ACRO466720250
Lanosterol 50%
Lanosterol
Formel: C₃₀H₅₀O
Smeltepunkt: 137°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00021108
CAS-nummer: 79-63-0
EINECS: 201-214-9

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow to green powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥ 50.0%
Specific optical rotation +60.0° to +72.0° (20°C, 589nm) (c=1, Toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR