Micro-spatula samplers, SteriWare
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8015R-170S 8015VB-170S 8015NB-170S 8015B-170S
300-0488EA 1730 DKK
300-0488 300-0489 300-0490 300-0491
Micro-spatula samplers, SteriWare
Prøvetagere Prøvetager til fast materiale
Disposable Micro-Spatulas are ideal for use with small amounts of powder and granule.
  • Ideal for sampling, dispensing, and measuring
  • Excellent for use in confined spaces
  • Available in two colours and multiple sizes
  • Easy to use
Sampling small amounts of powder or granule is made easy with the SteriWare Micro-Spatula. This single-use spatula is excellent when needing to take samples from confined spaces, and it eliminates the need for costly cleaning and cleaning validation.