VWR®, Density Standards, Certified Reference Materials, Referenced at 20 °C
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
99639.600EA 640 DKK
99639.600 99640.600 99641.600 99642.600 99649.600 99651.600 99652.600
VWR®, Density Standards, Certified Reference Materials, Referenced at 20 °C
Standarder Densitetsstandarder
VWR® ISO 17025 / ISO 17034 dual certified density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature.
  • ISO 17025 / ISO 17034 certified under UKAS accreditation
  • Density tested in strict accordance with ASTM D1480, a primary method for density determination
  • Density standards range between 0,6654 - 1,6300 g/ml
  • Ensures full compliance to ASTM and IP test method protocols
  • Shelf life between 12 / 24 months