MPC series couplings, medical grade
Leverandør: CPC COLDER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MPC17002T03 MPC17002T03B25 MPC17004T03B25 MPC17006T39B25
CPCCMPC17002T03EA 8060 DKK
CPCCMPC17002T03 CPCCMPC17002T03B25 CPCCMPC17004T03B25 CPCCMPC17006T39B25
MPC series couplings, medical grade
Fittings og stik
MPC Series couplings add ease of use and security to your most critical fluid handling applications.
  • Easy to operate
  • BSE/TSE free materials
  • Ergonomic thumb latch
  • Sterilisable by autoclave, Et0, e-beam or gamma
These couplings provide optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Operating temperature: −40 to 121 °C (for polycarbonate) and −40 to 149 °C (for polysulphone)
Certificeringer: USP Class VI, ADCF certified and meets biocompatibility requirements.