Safety goggles, GoggleGear™ 6000 series
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
GG6001SGAF-BLK GG6001SGAF-GRN GG6001SGAF-RED GG6001NSGAFBLU
MMMAGG6001SGAF-BLKEA 318 DKK
MMMAGG6001SGAF-BLK MMMAGG6001SGAF-GRN MMMAGG6001SGAF-RED MMMAGG6001NSGAFBLU
Safety goggles, GoggleGear™ 6000 series
Beskyttelsesbriller
GoggleGear™ safety goggles 6000 series features a large profile with indirect ventilation for air circulation with Scotchgard™ anti-fog and anti-scratch coating.
  • Large profile safety goggles provide enough space to fit over most prescription glasses
  • Indirect airflow to help against fogging
  • Premium spherical curved lens design for optimal horizontal and vertical field of vision
  • Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties
  • Pivoting ratchet for convenient personal adjustment
  • 2C-1.2 3M 1 BT K N lens marking
  • Wide flexible nose area to fit well with half-mask respirators
These large profile safety goggles feature indirect ventilation to allow air to circulate and help reduce fogging. The polycarbonate lenses feature a Scotchgard™ anti-fog and anti-scratch coating for superior anti-fog protection and increased durability that resists fogging for longer than traditional anti-fog coatings, even after up to 25 washings with water. Available in a range of shroud colours.
Blue goggles with neoprene headband can be sterilised up to 40x by autoclaving at 121 °C.